Learn More
Invitrogen™ PIGT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113837
Description
Immunogen sequence: PLPSGDVAAT FQFRTRWDSE LQREGVSHYR LFPKALGQLI SKYSLRELHL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
PIGT is a protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is an essential component of the multisubunit enzyme, GPI transamidase. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins. This gene encodes a protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is an essential component of the multisubunit enzyme, GPI transamidase. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins.
Specifications
| PIGT | |
| Polyclonal | |
| Unconjugated | |
| PIGT | |
| 2510012P17Rik; 4930534E15Rik; CGI-06; FLJ41596; GPI transamidase component PIG-T; GPI transamidase subunit; MCAHS3; MGC8909; NDAP; Ndap7; Neuronal development-associated protein 7; neurotrophin-regulated neuronal development-associated protein; phosphatidylinositol glycan anchor biosynthesis class T; phosphatidylinositol glycan anchor biosynthesis, class T; phosphatidylinositol glycan, class T; phosphatidylinositol-glycan biosynthesis class T protein; PIGT; PNH2; PSEC0163; si:ch211-153a8.2; UNQ716/PRO1379 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51604 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q969N2 | |
| PIGT | |
| Synthetic peptide directed towards the N terminal region of human PIGT (aa 38-87). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.