Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGU Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561313 Supplier Invitrogen™ Supplier No. PA561313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAEFISERVE VVSPLSSWKR VVEGLSLLDL GVSPYSGAVF HETPL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 87%.

Component of the GPI transamidase complex. May be involved in the recognition of either the GPI attachment signal or the lipid portion of GPI.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGU
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGU
Gene Accession No. Q9H490
Gene Alias 5430426F17Rik; BC028278; CDC91 (cell division cycle 91, S. cerevisiae, homolog)-like 1; CDC91 cell division cycle 91-like 1; CDC91L1; cell division cycle 91-like 1; cell division cycle 91-like 1 protein; cell division cycle protein 91-like 1; GAB1; GPI transamidase component PIG-U; GPI transamidase subunit; liver regeneration-related protein LRRGT00059; phosphatidylinositol glycan anchor biosynthesis class U; phosphatidylinositol glycan anchor biosynthesis class U protein; phosphatidylinositol glycan anchor biosynthesis, class U; phosphatidylinositol-glycan biosynthesis, class U protein; Pigu; Protein CDC91-like 1; PSEC0205; sakura; UNQ3055/PRO9875
Gene Symbols PIGU
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGU. Recombinant protein control fragment (Product #RP-99352).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.