Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598884
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598884 Supplier Invitrogen™ Supplier No. PA598884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIGV Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Glycosylphosphatidylinositol (GPI) is a complex glycolipid that anchors many proteins to the cell surface. The biosynthetic pathway of GPI is mediated by sequential addition of sugars and other components to phosphatidylinositol. PIGV adds the second mannose to the GPI core.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGV
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PIGV
Gene Accession No. Q9NUD9
Gene Alias B330013B03; D430024F16Rik; dol-P-Man dependent GPI mannosyltransferase; FLJ20477; GPI mannosyltransferase 2; GPI mannosyltransferase II; GPI-MT-II; HPMRS1; LOW QUALITY PROTEIN: GPI mannosyltransferase 2; phosphatidylinositol glycan anchor biosynthesis class V; phosphatidylinositol glycan anchor biosynthesis, class V; phosphatidylinositol glycan class V; phosphatidylinositol glycan, class V; phosphatidylinositol-glycan biosynthesis class V protein; Pigv; PIG-V; RGD1309526; Ybr004c homolog
Gene Symbols PIGV
Host Species Rabbit
Immunogen Recombinant Human GPI mannosyltransferase 2 protein (400-469AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55650
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.