Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGW Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537196
Description
This antibody detects endogenous protein at a molecular weight of 57 kDa. Purity is >95% by SDS-PAGE.
Phosphatidylinositol-glycans (PIGs) are multi-pass transmembrane proteins that localize to the endoplasmic reticulum. PIGs exhibit various functions, but all are crucial for the biosynthesis of the glycosylphosphatidylinositol (GPI)-anchor. Some PIG proteins are components of the GPI transamidase complex and play a role in the recognition of either the GPI attachment signal or the lipid portion of GPI. Other PIGs belong to the glycosyltransferase complex and function in the transfer of N-acetylglucosamine (GlcNAc) to phosphatidylinositol (PI). A variety of other PIGs play distinct roles in GPI synthesis including mannosylation of the GPI-anchor. PIG-W (Phosphatidylinositol-glycan biosynthesis class W protein) is a 504 amino acid multi-pass membrane protein that functions in the third step of GPI biosynthesis and acylates the inositol ring of phosphatidylinositol.
Specifications
PIGW | |
Polyclonal | |
Unconjugated | |
PIGW | |
2610044A17Rik; Gwt1; HPMRS5; phosphatidylinositol glycan anchor biosynthesis class W; phosphatidylinositol glycan anchor biosynthesis, class W; phosphatidylinositol glycan, class W; phosphatidylinositol-glycan biosynthesis class W protein; Pigw; PIG-W | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284098, 378774, 70325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7TSN4, Q7Z7B1, Q8C398 | |
PIGW | |
A synthetic peptide corresponding to residues in human PIGW. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction