Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556134 Supplier Invitrogen™ Supplier No. PA556134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MCSEIILRQE VLKDGFHRDL LIKVKFGESI EDLHTCRLLI KQDIPAGLYV DPYELASLRE RNITEAVMVS ENFDIEAPNY LSKESEVLIY ARRDSQCIDC FQAFLPVHCR YHRPHSEDGE ASIVVNNPDL LMFCD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Essential component of glycosylphosphatidylinositol-mannosyltransferase 1 which transfers the first of the 4 mannoses in the GPI-anchor precursors during GPI-anchor biosynthesis. Probably acts by stabilizing the mannosyltransferase PIGM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGX
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGX
Gene Accession No. Q8TBF5
Gene Alias 2010319C14Rik; GPI-mannosyltransferase subunit; phosphatidylinositol glycan anchor biosynthesis class X; phosphatidylinositol glycan anchor biosynthesis, class X; phosphatidylinositol glycan, class X; phosphatidylinositol-glycan biosynthesis class X protein; PIGX; PIG-X
Gene Symbols PIGX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGX. Recombinant protein control fragment (Product #RP-90387).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.