Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113251
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113251 Supplier Invitrogen™ Supplier No. PA5113251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIGX Polyclonal Antibody for Western Blot, IHC (P), ELISA

Essential component of glycosylphosphatidylinositol-mannosyltransferase 1 which transfers the first of the 4 mannoses in the GPI-anchor precursors during GPI-anchor biosynthesis. Probably acts by stabilizing the mannosyltransferase PIGM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGX
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PIGX
Gene Accession No. Q8TBF5, Q99LV7
Gene Alias 2010319C14Rik; GPI-mannosyltransferase subunit; phosphatidylinositol glycan anchor biosynthesis class X; phosphatidylinositol glycan anchor biosynthesis, class X; phosphatidylinositol glycan, class X; phosphatidylinositol-glycan biosynthesis class X protein; PIGX; PIG-X
Gene Symbols PIGX
Host Species Rabbit
Immunogen Recombinant Human Phosphatidylinositol-glycan biosynthesis class X protein (42-230AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54965, 72084
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.