Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520651
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520651 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520651 Supplier Invitrogen™ Supplier No. PA520651
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Despite its predicted molecular weight, PIG-Y often migrates at 28-30kDa in SDS-PAGE. A suggested positive control is A-20 cell lysate. PA5-20651 can be used with blocking peptide PEP-0771.

Glycosylphosphatidylinositol (GPI) lipid anchoring is an important post-translational modification of proteins that takes place in the endoplasmic reticulum. The synthesis of GPI is initiated by GPI-N-acetylglucosaminyltransferase (GPI-GnT), a complex of proteins including PIG-A, PIG-H, PIG-C, GPI1, and DPM2. PIG-Y, the mammalian homolog to yeast Eri1p, is also thought to be involved in the biosynthesis of GPI. The PIG-Y gene encodes two proteins, one of which arises from leaky scanning of the mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGY
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PIGY
Gene Accession No. P0C1P0, Q3MUY2
Gene Alias 1810008A14Rik; HPMRS6; phosphatidylinositol glycan anchor biosynthesis class Y; phosphatidylinositol glycan anchor biosynthesis, class Y-like; phosphatidylinositol N-acetylglucosaminyltransferase subunit Y; Phosphatidylinositol-glycan biosynthesis class Y protein; Pigy; PIG-Y; Pigyl; PREY; protein preY
Gene Symbols PIGY, Pigyl
Host Species Rabbit
Immunogen A 16 amino acid peptide near the carboxy terminus of human PIG-Y.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66268, 84992
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.