Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563595 Supplier Invitrogen™ Supplier No. PA563595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSWYFSSPAT SRNLVLTPVN FLHYNLNPQN LARHGTHARL THL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 81%.

The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells that serves to anchor proteins to the cell surface. This gene encodes a protein that is localized to the endoplasmic reticulum, and is involved in GPI anchor biosynthesis. As shown for the yeast homolog, which is a member of a family of dolichol-phosphate-mannose (Dol-P-Man)-dependent mannosyltransferases, this protein can also add a side-branching fourth mannose to GPI precursors during the assembly of GPI anchors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGZ
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGZ
Gene Accession No. Q86VD9
Gene Alias dol-P-Man dependent GPI mannosyltransferase; F630022B06Rik; GPI mannosyltransferase 4; GPI mannosyltransferase IV; GPI-MT-IV; hSMP3; LOW QUALITY PROTEIN: GPI mannosyltransferase 4; phosphatidylinositol glycan anchor biosynthesis class Z; phosphatidylinositol glycan anchor biosynthesis, class Z; phosphatidylinositol glycan, class Z; phosphatidylinositol glycan, class Z-like 1; phosphatidylinositol-glycan biosynthesis class Z protein; Pigz; PIG-Z; Pigzl1; RGD1307107; SMP3; SMP3 homolog; SMP3 mannosyltransferase
Gene Symbols PIGZ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGZ. Recombinant protein control fragment (Product #RP-101221).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80235
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.