Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGZ Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569106
Description
This target displays homology in the following species: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Yeast: 80%.
The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells that serves to anchor proteins to the cell surface. This gene encodes a protein that is localized to the endoplasmic reticulum, and is involved in GPI anchor biosynthesis. As shown for the yeast homolog, which is a member of a family of dolichol-phosphate-mannose (Dol-P-Man)-dependent mannosyltransferases, this protein can also add a side-branching fourth mannose to GPI precursors during the assembly of GPI anchors.
Specifications
PIGZ | |
Polyclonal | |
Unconjugated | |
PIGZ | |
dol-P-Man dependent GPI mannosyltransferase; F630022B06Rik; GPI mannosyltransferase 4; GPI mannosyltransferase IV; GPI-MT-IV; hSMP3; LOW QUALITY PROTEIN: GPI mannosyltransferase 4; phosphatidylinositol glycan anchor biosynthesis class Z; phosphatidylinositol glycan anchor biosynthesis, class Z; phosphatidylinositol glycan, class Z; phosphatidylinositol glycan, class Z-like 1; phosphatidylinositol-glycan biosynthesis class Z protein; Pigz; PIG-Z; Pigzl1; RGD1307107; SMP3; SMP3 homolog; SMP3 mannosyltransferase | |
Rabbit | |
Affinity chromatography | |
RUO | |
80235 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86VD9 | |
PIGZ | |
synthetic peptide directed towards the N terminal region of human PIGZ. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction