Learn More
Invitrogen™ PIGZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589818
Description
Immunogen sequence: LVPGLEYLEQ VVHAPVLPST PTHYTLLFTH TYMPPRHLLH LPGLGAPVEV VDMGGTEDWA LCQTLKSFTR QPACQVAGGP WLCRLFVVTP GTTRRAVEKC SFPFKNETLL FPHLTLEDPP ALSSLLSGAW RDHLSLHIVE LGEET; Positive Samples: U-87MG; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells that serves to anchor proteins to the cell surface. This gene encodes a protein that is localized to the endoplasmic reticulum, and is involved in GPI anchor biosynthesis. As shown for the yeast homolog, which is a member of a family of dolichol-phosphate-mannose (Dol-P-Man)-dependent mannosyltransferases, this protein can also add a side-branching fourth mannose to GPI precursors during the assembly of GPI anchors.
Specifications
| PIGZ | |
| Polyclonal | |
| Unconjugated | |
| PIGZ | |
| dol-P-Man dependent GPI mannosyltransferase; F630022B06Rik; GPI mannosyltransferase 4; GPI mannosyltransferase IV; GPI-MT-IV; hSMP3; LOW QUALITY PROTEIN: GPI mannosyltransferase 4; phosphatidylinositol glycan anchor biosynthesis class Z; phosphatidylinositol glycan anchor biosynthesis, class Z; phosphatidylinositol glycan, class Z; phosphatidylinositol glycan, class Z-like 1; phosphatidylinositol-glycan biosynthesis class Z protein; Pigz; PIG-Z; Pigzl1; RGD1307107; SMP3; SMP3 homolog; SMP3 mannosyltransferase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80235 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q86VD9 | |
| PIGZ | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 435-579 of human PIGZ (NP_0794392). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.