Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIH1D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100302
Description
Antibody detects endogenous levels of total PIH1D1.
Involved in the assembly of C/D box small nucleolar ribonucleoprotein (snoRNP) particles (PubMed:17636026). Recruits the SWI/SNF complex to the core promoter of rRNA genes and enhances pre-rRNA transcription (PubMed:22368283, PubMed:24036451). Mediates interaction of TELO2 with the R2TP complex which is necessary for the stability of MTOR and SMG1 (PubMed:20864032). Positively regulates the assembly and activity of the mTORC1 complex (PubMed:24036451).
Specifications
PIH1D1 | |
Polyclonal | |
Unconjugated | |
PIH1D1 | |
1110061L23Rik; 4933413A04Rik; NOP17; Nucleolar protein 17 homolog; PIH1 domain containing 1; PIH1 domain-containing protein 1; PIH1D1; RGD1309809 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
292898, 55011, 68845 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q4V7F5, Q9CQJ2, Q9NWS0 | |
PIH1D1 | |
A synthesized peptide derived from human PIH1D1(Accession Q9NWS0), corresponding to amino acid residues E84-N134. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction