Learn More
Invitrogen™ PIH1D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589383
Description
Immunogen sequence: MANPKLLGMG LSEAEAIGAD SARFEELLLQ ASKELQQAQT TRPESTQIQP QPGFCIKTNS SEGKVFINIC HSPSIPPPAD VTEEELLQML EEDQAGFRIP MSLGEPHAEL DAKGQGCTAY DVAVNSDFYR RMQNSDFLRE LVITIAREGL EDKYNLQLNP EWRMMKNRPF MGSISQQNIR SEQRPRIQEL GDLYTPAPGR AESGPEKPHL NLWLEAPDLL LAEVDLPKLD GALGLSLEIG ENRLVMGGPQ QLYHLDAYIP LQINSHESKA AFHRKRKQLM VAMPLLPVPS; Positive Samples: A-549, U-87MG, HeLa, A-431.
Involved in the assembly of C/D box small nucleolar ribonucleoprotein (snoRNP) particles (PubMed:17636026). Recruits the SWI/SNF complex to the core promoter of rRNA genes and enhances pre-rRNA transcription (PubMed:22368283, PubMed:24036451). Mediates interaction of TELO2 with the R2TP complex which is necessary for the stability of MTOR and SMG1 (PubMed:20864032). Positively regulates the assembly and activity of the mTORC1 complex (PubMed:24036451).
Specifications
PIH1D1 | |
Polyclonal | |
Unconjugated | |
PIH1D1 | |
1110061L23Rik; 4933413A04Rik; NOP17; Nucleolar protein 17 homolog; PIH1 domain containing 1; PIH1 domain-containing protein 1; PIH1D1; RGD1309809 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55011 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.35 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9NWS0 | |
PIH1D1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human PIH1D1 (NP_0603861). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.