Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIH1D2 Antibody (OTI4A10), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273403AF350
Description
PIH1D2 Monoclonal specifically detects PIH1D2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
PIH1D2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
PIH1D2 | |
Recombinant protein fragment corresponding to amino acids 1-315 of human PIH1D2 (NP_620144) produced in E.coli. | |
0.1 mL | |
120379.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4A10 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
PIH1 domain containing 2, PIH1 domain-containing protein 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction