Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIH1D2 Antibody (OTI4A10), Janelia Fluor™ 585, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273403JF585
Description
PIH1D2 Monoclonal antibody specifically detects PIH1D2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOFSpecifications
| PIH1D2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 120379 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI4A10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| PIH1 domain containing 2, PIH1 domain-containing protein 2 | |
| Human recombinant protein fragment corresponding to amino acids 1-315 of human PIH1D2(NP_620144) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction