Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIH1D2 Antibody (OTI4A10), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273403MFV610
Description
PIH1D2 Monoclonal antibody specifically detects PIH1D2 in Human samples. It is validated for Flow CytometrySpecifications
PIH1D2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
120379 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI4A10 | |
Flow Cytometry | |
PIH1 domain containing 2, PIH1 domain-containing protein 2 | |
Human recombinant protein fragment corresponding to amino acids 1-315 of human PIH1D2(NP_620144) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction