Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIH1D2 Monoclonal Antibody (OTI4A10)
GREENER_CHOICE

Catalog No. PIMA525429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525429 Supplier Invitrogen™ Supplier No. MA525429
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIH1D2 Monoclonal Antibody for Western Blot, IHC (P), Flow

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIH1D2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4A10
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIH1D2
Gene Accession No. Q8WWB5
Gene Alias 2700059L22Rik; PIH1 domain containing 2; PIH1 domain-containing protein 2; Pih1d2
Gene Symbols PIH1D2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-315 of PIH1D2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120379
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.