Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIH1D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571511
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571511 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571511 Supplier Invitrogen™ Supplier No. PA571511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIH1D2 Polyclonal Antibody for Western Blot, IHC (P)

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIH1D2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PIH1D2
Gene Accession No. Q8CHR9, Q8WWB5
Gene Alias 2700059L22Rik; PIH1 domain containing 2; PIH1 domain-containing protein 2; Pih1d2
Gene Symbols PIH1D2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 220-249 amino acids from the C-terminal region of human PIH1D2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120379, 72614
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.