Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIH1D3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734852 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4852 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4852 Supplier Proteintech Group Inc Supplier No. 253091AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays a role in cytoplasmic pre-assembly of axonemal dynein.

Specifications

Antigen PIH1D3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PIH1D3
Gene Accession No. Q9NQM4
Gene Alias CXorf41, PIH1D3, Sarcoma antigen NY SAR 97
Gene Symbols PIH1D3
Host Species Rabbit
Immunogen PIH1D3 Fusion Protein Ag15039
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139212
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.