Learn More
PIK3AP1 Mouse anti-Human, Rat, Clone: OTI6H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501764

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.Specifications
PIK3AP1 | |
Monoclonal | |
Unconjugated | |
PIK3AP1 | |
BCAP | |
Mouse | |
Affinity Chromatography | |
RUO | |
118788, 294048 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI6H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6ZUJ8 | |
Pik3ap1 | |
Full length human recombit protein of human PIK3AP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.