Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3AP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558397 Supplier Invitrogen™ Supplier No. PA558397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EWQLNQKKRS ESFRFQQENL KRLRDSITRR QREKQKSGKQ TDLEITVPIR HSQHLPAKVE FGVYESGPRK SVIPPRTELR RGDWK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.

PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3AP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIK3AP1
Gene Accession No. Q6ZUJ8
Gene Alias 1810044J04Rik; B cell adaptor protein; B cell phosphoinositide 3-kinase adaptor; BCAP; B-cell adapter for phosphoinositide 3-kinase; B-cell phosphoinositide 3-kinase adapter protein 1; FLJ35564; Phosphoinositide 3-kinase adapter protein 1; phosphoinositide-3-kinase; phosphoinositide-3-kinase adaptor protein 1; PIK3AP1; RP11-34E5.3
Gene Symbols PIK3AP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCAP. Recombinant protein control fragment (Product #RP-102833).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118788
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.