Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIK3AP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | PIK3AP1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119342
![]() |
Invitrogen™
PA5119342 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121683
|
Invitrogen™
PA5121683 |
20 μL |
Each for $209.50
|
|
|||||
Description
PIK3AP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.Specifications
PIK3AP1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
PIK3AP1 | |
1810044J04Rik; B cell adaptor protein; B cell phosphoinositide 3-kinase adaptor; BCAP; B-cell adapter for phosphoinositide 3-kinase; B-cell phosphoinositide 3-kinase adapter protein 1; FLJ35564; Phosphoinositide 3-kinase adapter protein 1; phosphoinositide-3-kinase; phosphoinositide-3-kinase adaptor protein 1; PIK3AP1; RP11-34E5.3 | |
PIK3AP1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q6ZUJ8, Q9EQ32 | |
118788, 83490 | |
Recombinant protein Phosphoinositide-3-Kinase Adaptor Protein 1. The antigen corresponds to amino acid range 489-730 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title