Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIK3AP1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547663
Description
This antibody detects endogenous human and mouse BCAP in Western blots. Reconstitute at 0.2 mg/mL in sterile PBS.
PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.
Specifications
PIK3AP1 | |
Polyclonal | |
Unconjugated | |
PIK3AP1 | |
1810044J04Rik; B cell adaptor protein; B cell phosphoinositide 3-kinase adaptor; BCAP; B-cell adapter for phosphoinositide 3-kinase; B-cell phosphoinositide 3-kinase adapter protein 1; FLJ35564; Phosphoinositide 3-kinase adapter protein 1; phosphoinositide-3-kinase; phosphoinositide-3-kinase adaptor protein 1; PIK3AP1; RP11-34E5.3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
118788, 83490 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6ZUJ8, Q9EQ32 | |
PIK3AP1 | |
E. coli-derived recombinant human BCAP Met462-Leu650. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction