Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3C2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18090825UL
Description
PIK3C2A Polyclonal specifically detects PIK3C2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PIK3C2A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
O00443 | |
PIK3C2A | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CGTKLLYLWTSSHTNSVPGTVTKKGYVMERIVLQVDFPSPAFDIIYTTPQVDRSIIQQHNLETLENDIKGKLLDILHKDSSLGLSKEDK | |
25ul | |
Signal Transduction | |
5286 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C2-containing phosphatidylinositol kinase, CPK, DKFZp686L193, EC 2.7.1, EC 2.7.1.154, MGC142218, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha, Phosphoinositide 3-kinase-C2-alpha, phosphoinositide-3-kinase, class 2, alpha polypeptide, PI3-K-C2(ALPHA), PI3-K-C2A, PI3K-C2alpha, PI3K-C2-alpha, PtdIns-3-kinase C2 subunit alpha | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction