Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3C2A Monoclonal Antibody (OTI3D9)
GREENER_CHOICE

Catalog No. PIMA526509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526509 Supplier Invitrogen™ Supplier No. MA526509
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIK3C2A Monoclonal Antibody for Western Blot, IHC (P)

The Class II PI3 kinases, including PIK3C2A, phosphorylate the 3 position of PI or PI4P. Though not as well studied as the Class I PI3 kinases, the Class II enyzmes have been linked to diverse, receptor-mediated cellular processes including insulin signaling, neuronal survival, and growth factor signaling. PI3-kinases play roles in signaling pathways involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. This protein contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases. C2 domains act as calcium-dependent phospholipid binding motifs that mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The PI3-kinase activity of this protein is not sensitive to nanomolar levels of the inhibitor wortmanin. This protein was shown to be able to be activated by insulin and may be involved in integrin-dependent signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3C2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIK3C2A
Gene Accession No. O00443
Gene Alias C2-containing phosphatidylinositol kinase; CPK; Cpk-m; DKFZp686L193; MGC142218; p170; phosphatidylinositol 3-kinase, C2 domain containing, alpha polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha; phosphoinositide 3-kinase-C2-alpha; phosphoinositide-3-kinase, class 2, alpha polypeptide; PI3KC2; PI3-K-C2(ALPHA); pi3kc2a; PI3-K-C2A; PI3K-C2alpha; PI3K-C2-alpha; Pik3c2a; ptdIns-3-kinase C2 subunit alpha
Gene Symbols PIK3C2A
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 230-560 of PIK3C2A produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5286
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.