Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3C2G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599834 Supplier Invitrogen™ Supplier No. PA599834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PIK3C2G.

PIK3C2G lipid kinase belongs to the phosphoinositide 3-kinase (PI3K) family which play major roles in signaling pathways and are involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. PIK3C2G contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases that act as calcium-dependent phospholipid binding motifs which mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The N-terminal region of PIK3C2G and the C-terminal C2 domain are required for enzymatic activity. The PI 3-kinase C2gamma gene encodes a 1,050 amino acid polypeptide with 36% identity to human PI 3-kinase C2alpha. Research indicates that PI 3-kinase C2gamma can block the growth of human colon cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3C2G
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PIK3C2G
Gene Accession No. O70167, O75747
Gene Alias C80387; phosphatidylinositol 3-kinase, C2 domain containing, gamma polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit gamma; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing gamma polypeptide; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit gamma; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 gamma; phosphoinositide 3-Kinase-C2-gamma; phosphoinositide-3-kinase, class 2, gamma polypeptide; PI 3-kinase C2gamma; PI3KC2gamma; PI3K-C2gamma; PI3K-C2-gamma; Pik3c2g; PTDINS-3-kinase C2 gamma; ptdIns-3-kinase C2 subunit gamma
Gene Symbols PIK3C2G
Host Species Rabbit
Immunogen A synthesized peptide derived from human PIK3C2G(Accession O75747), corresponding to amino acid residues S1362-I1412.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18705, 5288
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.