Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3CA Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298814B
Description
PIK3CA Polyclonal specifically detects PIK3CA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PIK3CA | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Biotin | |
EC 2.7.1, EC 2.7.1.153, MGC142161, MGC142163, p110-alpha, Phosphatidylinositol 3-Kinase catalytic subunit alpha, phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha, phosphatidylinositol 3-kinase, catalytic, alpha polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform, phosphoinositide-3-kinase, catalytic, alpha polypeptide, PI3K, PI3K-alpha, PI3-kinase p110 subunit alpha, PI3-kinase subunit alpha, PtdIns-3-kinase p110, PtdIns-3-kinase subunit alpha, PtdIns-3-kinase subunit p110-alpha | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PIK3CA. | |
0.1 mL | |
Breast Cancer, Cancer, Diabetes Research, mTOR Pathway, Oncogenes | |
5290 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction