Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PIK3CA E545K/PIK3R1 (p110 alpha E545K/p85 alpha), His Tag Recombinant Protein

Catalog No. A32909
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
20 μg

Recombinant Protein

PIK3CA E542K/PIK3R1 (p110 alpha E542K/p85 alpha) lipid kinase is comprised of a 110 kDa catalytic subunit and an 85 kDa regulatory subunit. A number of isoforms of the 110 kDa catalytic subunit and the 85 kDa regulatory subunit exist in cells. PIK3CA is frequently mutated or amplified in a variety of cancers including ovarian and colon and this kinase is one of the PI3K mutants. PIK3CA gene copy number is increased in over 30% of ovarian cancers and this leads to increased PI3-kinase activity. Furthermore, the activity of PIK3CA is essential for vascular development and inactivation of PIK3CA leads to severe defects in angiogenic sprouting and vascular remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27986, P42336
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM sodium phosphate with 0.1 mM PMSF, 0.25 mM DTT, 150 mM Imidazole, 25% glycerol, 300 mM NaCl and no preservative; pH 7
Gene ID (Entrez) 5290, 5295
Molecular Weight (g/mol) 125.4-84.7 kDa
Name Human PIK3CA E545K/PIK3R1 (p110 alpha E545K/p85 alpha), His Tag
Quantity 100 μg
Gene Alias PIK3CA; PIK3R1
Common Name PIK3CA (E545K Mutant)/PIK3R1
KinaseFamily PI3 Kinase
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active, Mutant
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.