Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3CA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587398 Supplier Invitrogen™ Supplier No. PA587398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa Immunogen sequence: RLCLSICSVK GRKGAKEEHC PLAWGNINLF DYTDTLVSGK MALNLWPVPH GLEDLLNPIG VTGSNPNKET PCLELEFDWF SSVVKFPDMS VIEEHANWSV SREAGFSYSH AGLSNRLARD NELRENDKEQ LKAISTRDPL SEITEQEKDF LWSHRHYCVT IPEILPKLLL SVKWNSRDEV AQMYCLVKDW PPIKPEQAME.

Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3CA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIK3CA
Gene Accession No. A0A0G2K344, P42336, P42337
Gene Alias 2.7.1.153; 6330412C24Rik; caPI3K; CLOVE; CWS5; EC:2.7.11.1; GRB1; MCAP; MCM; MCMTC; MGC142161; MGC142163; p110; p110alpha; p110-alpha; phophatidylinositol 3-kinase; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha; phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3K; PI3K alpha; PI3Kalpha; PI3K-alpha; pi3kca; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; Pik3ca; PIK3R1; pk3ca; ptdIns-3-kinase p110; ptdIns-3-kinase subunit alpha; ptdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA
Gene Symbols PIK3CA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 401-600 of human PIK3CA (NP_0062092).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170911, 18706, 5290
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.