Learn More
Invitrogen™ PIK3CB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120062
Description
Positive test controls include: MCF7, Mouse brain. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: KVKTKKSTKT INPSKYQTIR KAGKVHYPVA WVNTMVFDFK GQLRTGDIIL HSWSSFPDEL EEMLNPMGTV QTNPYTENAT ALHVKFPENK KQPYYYPPFD KIIEKAAEIA SSDSANVSSR GGKKFLPVLK EILDRDPLSQ LCENEMDLIW TLRQDCREIF PQSLPKLLLS IKWNKLEDVA QLQALLQIWP K.
PIK3CB (phosphatyidylinositol 4,5-biphosphate 3-kinase catalyic subunit beta isoform) is a phosphoinositide-3-kinase that phosphorylates PtdIns (phosphatidylinositol), PtdIns4P (phosphatidylinositol 4-phosphate), and PtdIns(4,5)P2 (phosphatidylinositol 4,5-biphosphate) to generate phosphatidylinositol 3,4,5-triphosphate (PIP3). PIP3 plays a key role in recruiting PH domain-containing proteins to the membrane and activating signaling cascades involved in cell growth, survival, proliferation, motility, and morphology.
Specifications
PIK3CB | |
Polyclonal | |
Unconjugated | |
PIK3CB | |
1110001J02Rik; AI447572; catalytic phosphatidylinositol 3-kinase beta; p110beta; phosphatidylinositol 3-kinase catalytic subunit beta isoform; phosphatidylinositol 3-kinase, catalytic subunit, beta isoform; phosphatidylinositol 3-kinase, catalytic, beta polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit beta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta; phosphoinositide-3-kinase, catalytic, beta polypeptide; PI3K; PI3Kbeta; PI3K-beta; pi3kcb; PI3-kinase p110 subunit beta; PI3-kinase subunit beta; PIK3C1; Pik3cb; PtdIns-3-kinase p110; ptdIns-3-kinase subunit beta; ptdIns-3-kinase subunit p110-beta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5291, 74769, 85243 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.69 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42338, Q8BTI9, Q9Z1L0 | |
PIK3CB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 410-600 of human PI3 Kinase p110 beta (NP_0062101). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.