Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3CB Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992092
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2092 25 μL
50-199-2091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2092 Supplier Rockland Immunochemicals Supplier No. 600401GT8S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human PIK3C beta protein. A BLAST analysis was used to suggest cross-reactivity with PI3K-beta based on a 100% homology from human sources with the immunizing sequence. Expect partial cross reactivity to PIK3C beta from mouse and rat (93% homology) sources. Cross-reactivity with PIK3C beta from other sources has not been determined.

PIK3CB (phosphatyidylinositol 4,5-biphosphate 3-kinase catalyic subunit beta isoform) is a phosphoinositide-3-kinase that phosphorylates PtdIns (phosphatidylinositol), PtdIns4P (phosphatidylinositol 4-phosphate), and PtdIns(4,5)P2 (phosphatidylinositol 4,5-biphosphate) to generate phosphatidylinositol 3,4,5-triphosphate (PIP3). PIP3 plays a key role in recruiting PH domain-containing proteins to the membrane and activating signaling cascades involved in cell growth, survival, proliferation, motility, and morphology.

Specifications

Antigen PIK3CB
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene PIK3CB
Gene Accession No. P42338
Gene Alias 1110001J02Rik; AI447572; catalytic phosphatidylinositol 3-kinase beta; p110beta; phosphatidylinositol 3-kinase catalytic subunit beta isoform; phosphatidylinositol 3-kinase, catalytic subunit, beta isoform; phosphatidylinositol 3-kinase, catalytic, beta polypeptide; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit beta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta; phosphoinositide-3-kinase, catalytic, beta polypeptide; PI3K; PI3KBETA; PI3K-beta; PI3-kinase p110 subunit beta; PI3-kinase subunit beta; PIK3C1; PIK3CB; ptdIns-3-kinase p110; ptdIns-3-kinase subunit beta; PtdIns-3-kinase subunit p110-beta
Gene Symbols PIK3CB
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5291
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.