Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3CB Recombinant Rabbit Monoclonal Antibody (2F1B7)
GREENER_CHOICE

Catalog No. MA568857
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA568857 20 μL
FEMA551436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA568857 Supplier Invitrogen™ Supplier No. MA568857
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KVKTKKSTKT INPSKYQTIR KAGKVHYPVA WVNTMVFDFK GQLRTGDIIL HSWSSFPDEL EEMLNPMGTV QTNPYTENAT ALHVKFPENK KQPYYYPPFD KIIEKAAEIA SSDSANVSSR GGKKFLPVLK EILDRDPLSQ LCENEMDLIW TLRQDCREIF PQSLPKLLLS IKWNKLEDVA QLQALLQIWP K.

PIK3CB (phosphatyidylinositol 4,5-biphosphate 3-kinase catalyic subunit beta isoform) is a phosphoinositide-3-kinase that phosphorylates PtdIns (phosphatidylinositol), PtdIns4P (phosphatidylinositol 4-phosphate), and PtdIns(4,5)P2 (phosphatidylinositol 4,5-biphosphate) to generate phosphatidylinositol 3,4,5-triphosphate (PIP3). PIP3 plays a key role in recruiting PH domain-containing proteins to the membrane and activating signaling cascades involved in cell growth, survival, proliferation, motility, and morphology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3CB
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2F1B7
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PIK3CB
Gene Accession No. P42338, Q8BTI9, Q9Z1L0
Gene Alias 1110001J02Rik; AI447572; catalytic phosphatidylinositol 3-kinase beta; p110beta; phosphatidylinositol 3-kinase catalytic subunit beta isoform; phosphatidylinositol 3-kinase, catalytic subunit, beta isoform; phosphatidylinositol 3-kinase, catalytic, beta polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit beta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta; phosphoinositide-3-kinase, catalytic, beta polypeptide; PI3K; PI3Kbeta; PI3K-beta; pi3kcb; PI3-kinase p110 subunit beta; PI3-kinase subunit beta; PIK3C1; Pik3cb; PtdIns-3-kinase p110; ptdIns-3-kinase subunit beta; ptdIns-3-kinase subunit p110-beta
Gene Symbols PIK3CB
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5291, 74769, 85243
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.