Learn More
PIK3CD Mouse anti-Human, Clone: OTI1F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801842

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifications
PIK3CD | |
Monoclonal | |
Unconjugated | |
PIK3CD | |
APDS, IMD14, P110DELTA, PI3K, p110D | |
Mouse | |
Affinity Chromatography | |
RUO | |
5293 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O00329 | |
PIK3CD | |
Human recombit protein fragment corresponding to amino acids 286-610 of human PIK3CD produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.