Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3CD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583748 Supplier Invitrogen™ Supplier No. PA583748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AECSRLLQIL ELGRHSECVH VTEEEQLQLR EILERRGSGE LYEHEKDLVW KLRHEVQEHF PEALARLLLV TKWN Highest antigen sequence indentity to the following orthologs: Rat - 34%, Mouse - 82%.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3CD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIK3CD
Gene Accession No. O00329
Gene Alias 2410099E07Rik; 2610208K16Rik; APDS; AW545373; catalytic phosphatidylinositol 3-kinase delta; IMD14; p100 delta; p110D; p110delta; phosphatidylinositol 3-kinase catalytic delta polypeptide; phosphatidylinositol 3-kinase catalytic subunit delta; phosphatidylinositol 3-kinase p110 delta; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta; phosphoinositide 3 kinase P110delta; phosphoinositide-3-kinase C; phosphoinositide-3-kinase, catalytic, delta polypeptide; phosphoinositide-3-kinase, catalytic, delta polypeptide variant p37delta; PI3K; PI3K delta; pi3kcd; PI3Kdelta; PI3K-delta; PI3-kinase p110 subunit delta; PI3-kinase subunit delta; PI3Kp110delta; PI3K-p110delta; PIK3CD; ptdIns-3-kinase subunit delta; ptdIns-3-kinase subunit p110-delta
Gene Symbols PIK3CD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIK3CD. Recombinant protein control fragment (Product #RP-102971).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5293
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.