Learn More
Invitrogen™ PIK3CG Polyclonal Antibody

Description
Positive test controls include: Jurkat, K-562, Mouse thymus. The target is usually found in the following locations: Cell membrane, Cytoplasm. Immunogen sequence: RSPGQIHLVQ RHPPSEESQA FQRQLTALIG YDVTDVSNVH DDELEFTRRG LVTPRMAEVA SRDPKLYAMH PWVTSKPLPE YLWKKIANNC IFIVIHRSTT SQTIKVSPDD TPGAILQSFF TKMAKKKSLM DIPESQSEQD FVLRVCGRDE YLVGETPIKN FQWVRHCLKN GEEIHVVLDT PPDPALDEVR KEEWPLVDDC TGVTGYHEQL TIH.
Specifications
Specifications
| Antigen | PIK3CG |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | PIK3CG |
| Gene Accession No. | P48736, Q9JHG7 |
| Gene Alias | 1-phosphatidylinositol 3-kinase; 5830428L06Rik; EGK_14026; p110gamma; p110-gamma; p120-PI3K; phosphatidylinositol 3 kinase gamma, p110 gamma; phosphatidylinositol 3-kinase catalytic 110-kD gamma; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit gamma; Phosphoinositide-3-kinase catalytic gamma polypeptide; phosphoinositide-3-kinase gamma catalytic subunit; phosphoinositide-3-kinase, catalytic, gamma polypeptide; PI3CG; PI3K; pi3kcg; Pi3kg1; PI3Kgamma; PI3K-gamma; PI3-kinase subunit gamma; PIK3; Pik3cg; ptdIns-3-kinase subunit gamma; ptdIns-3-kinase subunit p110-gamma; Serine/threonine protein kinase PIK3CG |
| Gene Symbols | PIK3CG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.