Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3IP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100172 Supplier Invitrogen™ Supplier No. PA5100172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PIK3IP1.

The exact function of PIK3IP1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3IP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pik3ip1
Gene Accession No. Q56A20, Q7TMJ8, Q96FE7
Gene Alias 1500004A08Rik; 5830455E04Rik; calcineurin-regulated kringle domain protein; calcineurin-regulated kringle domain-containing protein; Crkd; HGFL; HGFL protein; hHGFL(S); Kringle domain-containing protein HGFL; phosphoinositide-3-kinase interacting protein 1; phosphoinositide-3-kinase-interacting protein 1; Pik3ip1; RGD1311203; zgc:66482
Gene Symbols Pik3ip1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PIK3IP1(Accession Q96FE7), corresponding to amino acid residues L2-A52.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113791, 216505, 305472
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.