Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIK3IP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113261
Description
PIK3IP1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
The exact function of PIK3IP1 remains unknown.
Specifications
| PIK3IP1 | |
| Polyclonal | |
| Unconjugated | |
| Pik3ip1 | |
| 1500004A08Rik; 5830455E04Rik; calcineurin-regulated kringle domain protein; calcineurin-regulated kringle domain-containing protein; Crkd; HGFL; HGFL protein; hHGFL(S); Kringle domain-containing protein HGFL; phosphoinositide-3-kinase interacting protein 1; phosphoinositide-3-kinase-interacting protein 1; Pik3ip1; RGD1311203; zgc:66482 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 113791, 216505 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.48 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q7TMJ8, Q96FE7 | |
| Pik3ip1 | |
| Recombinant Human Phosphoinositide-3-kinase-interacting protein 1 protein (22-168AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction