Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120556 Supplier Invitrogen™ Supplier No. PA5120556
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Mouse kidney, Rat brain. Immunogen sequence: GLTLPDLPEQ FSPPDVAPPL LVKLVEAIER TGLDSESHYR PELPAPRTDW SLSDVDQWDT AALADGIKSF LLALPAPLVT PEASAEARRA LREAAGPVGP ALEPPTLPLH RALTLRFLLQ HLGRVARRAP ALGPAVRALG ATFGPLLLRA PPPPSSPPPG GAPDGSEPSP DFPALLVEKL LQEHLEEQEV A.

Binds to activated (phosphorylated) protein-tyrosine kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3R2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PIK3R2
Gene Accession No. O00459, O08908, Q63788
Gene Alias MPPH; MPPH1; p85; P85B; p85beta; p85-BETA; phosphatidylinositol 3-kinase; phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase regulartory subunit polypeptide 2; phosphatidylinositol 3-kinase regulatory subunit beta; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphoinositide-3-kinase regulatory subunit 2; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta); phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta); PI3K; PI3K regulatory subunit beta; PI3-kinase p85 subunit beta; PI3-kinase regulatory subunit beta; PI3-kinase subunit p85-beta; PIK3R2; ptdIns-3-kinase p85-beta; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit p85-beta
Gene Symbols PIK3R2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 108-298 of human PI3 Kinase p85 beta (NP_005018.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18709, 29741, 5296
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.