Learn More
PIK3R2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302594A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Binds to activated (phosphorylated) protein-tyrosine kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane.Specifications
PIK3R2 | |
Polyclonal | |
Unconjugated | |
PIK3R2 | |
P85B, p85, p85-BETA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5296 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O00459 | |
PIK3R2 | |
Between 125 and 175 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.