Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIK3R4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120397 Supplier Invitrogen™ Supplier No. PA5120397
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Raji, Mouse thymus, Rat lung. The target is usually found in the following locations: Cytoplasmic vesicle, Late endosome, Lipid-anchor, Membrane, autophagosome. Immunogen sequence: MGNQLAGIAP SQILSVESYF SDIHDFEYDK SLGSTRFFKV ARAKHREGLV VVKVFAIQDP TLPLTSYKQE LEELKIRLNS AQNCLPFQKA SEKASEKAAM LFRQYVRDNL YDRISTRPFL NNIEKRWIAF QILTAVDQAH KSGVRHGDIK TENVMVTSWN WVLLTDFASF KPTYLPEDNP ADFNYFFDTS RRRTCYIAPE RFVDGGMFAT ELEYMRDPST PLVDLNSNQR TRGELKRAMD IFSAGCVIAE LFTEGVPLFD LSQLLAYRNG HFFPEQVLNK IEDHSIRELV TQMIHREPDK RLEAEDYLKQ QRGNAFPEIF YTFLQPYMAQ.

Regulatory subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate; different complex forms are believed to play a role in multiple membrane trafficking pathways: PI3KC3-C1 is involved in initiation of autophagosomes and PI3KC3-C2 in maturation of autophagosomes and endocytosis. Involved in regulation of degradative endocytic trafficking and cytokinesis, probably in the context of PI3KC3-C2 (PubMed:20643123). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3R4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PIK3R4
Gene Accession No. P0C0R5, Q8VD65, Q99570
Gene Alias 2210010O15Rik; C730038E05Rik; C85833; D9Ertd418e; p150; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 4, p150; phosphatidylinositol 3-kinase-associated p150; phosphoinositide 3-kinase adaptor protein; Phosphoinositide 3-kinase regulatory subunit 4; phosphoinositide-3-kinase regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4, p150; PI3-Kinase p150; PI3-kinase p150 subunit; PI3-kinase regulatory subunit 4; PI3-kinase VPS15; PIK3R4; VPS15
Gene Symbols PIK3R4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-330 of human PIK3R4/VPS15 (NP_055417.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30849, 363131, 75669
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.