Learn More
Invitrogen™ PIK3R4 Polyclonal Antibody

Description
Recommended positive controls: T-47D cell lysate.
Specifications
Specifications
| Antigen | PIK3R4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | PIK3R4 |
| Gene Accession No. | Q99570 |
| Gene Alias | 2210010O15Rik; C730038E05Rik; C85833; D9Ertd418e; p150; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 4, p150; phosphatidylinositol 3-kinase-associated p150; phosphoinositide 3-kinase adaptor protein; Phosphoinositide 3-kinase regulatory subunit 4; phosphoinositide-3-kinase regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4, p150; PI3-Kinase p150; PI3-kinase p150 subunit; PI3-kinase regulatory subunit 4; PI3-kinase VPS15; PIK3R4; VPS15 |
| Gene Symbols | PIK3R4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.