Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3R5 Antibody (OTI4G9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273408AF532
Description
PIK3R5 Monoclonal specifically detects PIK3R5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PIK3R5 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
PIK3R5 | |
Full length recombinant protein of human PIK3R5 (NP_055123) produced in HEK293T cells. | |
0.1 mL | |
23533.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4G9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
F730038I15Rik, FOAP-2, p101, P101-PI3K, Phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit, phosphoinositide-3-kinase, regulatory subunit 5, phosphoinositide-3-kinase, regulatory subunit, polypeptide p101, PI3-kinase p101 subunit, PI3-kinase regulatory subunit 5, PtdIns-3-kinase regulatory subunit, regulatory subunit 5, p101 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction