Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3R5 Antibody (OTI4G9), Janelia Fluor™ 525, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273408JF525
Description
PIK3R5 Monoclonal antibody specifically detects PIK3R5 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
PIK3R5 | |
Monoclonal | |
Janelia Fluor 525 | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
23533 | |
Store at 4°C in the dark. | |
IgG2b |
Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
OTI4G9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
F730038I15Rik, FOAP-2, p101, P101-PI3K, Phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit, phosphoinositide-3-kinase, regulatory subunit 5, phosphoinositide-3-kinase, regulatory subunit, polypeptide p101, PI3-kinase p101 subunit, PI3-kinase regulatory subunit 5, PtdIns-3-kinase regulatory subunit, regulatory subunit 5, p101 | |
Full length human recombinant protein of human PIK3R5(NP_055123) produced in HEk293T cell. | |
0.1 mL | |
Primary | |
Canine, Human, Monkey, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction