Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3R5 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
SDP

Catalog No. 501676236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-6236 Supplier OriGene Supplier No. TA505828
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Receptor-regulated class I phosphoinositide 3-kinases (PI3Ks) phosphorylate the membrane lipid phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) to PtdIns(3,4,5)P3, which in turn recruits and activates cytosolic effectors involved in proliferation, survival, or chemotaxis. PIK3R5 is a PI3K regulatory subunit.

Specifications

Antigen PIK3R5
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PIK3R5
Gene Accession No. Q8WYR1
Gene Alias F730038I15Rik, FOAP-2, P101-PI3K, p101
Gene Symbols PIK3R5
Host Species Mouse
Immunogen Full length human recombit protein of human PIK3R5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23533
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.