Learn More
Invitrogen™ PIK3R5 Monoclonal Antibody (OTI5B1)

Description
PIK3R5 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | PIK3R5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI5B1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PIK3R5 |
| Gene Accession No. | Q5SW28, Q8WYR1 |
| Gene Alias | AV230647; F730038I15Rik; FOAP-2; p101; p101-PI3K; phosphatidylinositol 4,5-bisphosphate 3-kinase regulatory subunit; phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit; phosphoinositide 3-kinase regulatory subunit 5; phosphoinositide-3-kinase regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5, p101; PI3-kinase p101 subunit; PI3-kinase regulatory subunit 5; Pik3r5; Protein FOAP-2; ptdIns-3-kinase p101; PtdIns-3-kinase regulatory subunit; RGD1563261 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.