Learn More
PIK3R5 Mouse anti-Human, Clone: OTI4C12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505827

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Receptor-regulated class I phosphoinositide 3-kinases (PI3Ks) phosphorylate the membrane lipid phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) to PtdIns(3,4,5)P3, which in turn recruits and activates cytosolic effectors involved in proliferation, survival, or chemotaxis. PIK3R5 is a PI3K regulatory subunit.Specifications
PIK3R5 | |
Monoclonal | |
Unconjugated | |
PIK3R5 | |
F730038I15Rik, FOAP-2, P101-PI3K, p101 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23533 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4C12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WYR1 | |
PIK3R5 | |
Full length human recombit protein of human PIK3R5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.