Learn More
Invitrogen™ PIK3R5 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PI3 kinase p101 protein.
Specifications
Specifications
| Antigen | PIK3R5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PIK3R5 |
| Gene Accession No. | Q5SW28, Q8WYR1 |
| Gene Alias | AV230647; F730038I15Rik; FOAP-2; p101; p101-PI3K; phosphatidylinositol 4,5-bisphosphate 3-kinase regulatory subunit; phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit; phosphoinositide 3-kinase regulatory subunit 5; phosphoinositide-3-kinase regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5, p101; PI3-kinase p101 subunit; PI3-kinase regulatory subunit 5; Pik3r5; Protein FOAP-2; ptdIns-3-kinase p101; PtdIns-3-kinase regulatory subunit; RGD1563261 |
| Gene Symbols | PIK3R5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.