Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIK3R6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554689
Description
Immunogen sequence: PAAPVTDHTC LNVNVTEVVK SSNLAGKSFS TVTNTFRTNN IQIQSRDQRL LTLSLDKDDQ RTFRDVVRFE VAPCPEPCSG AQKSKAPWLN LHGQQEVEAI KAKPKP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 67%.
p87PIKAP (p87 PI3K adapter protein), also known as PIK3R6 (phosphoinositide-3-kinase, regulatory subunit 6), p84 or HsT41028, is a 754 amino acid cytoplasmic protein that functions as a regulatory subunit of the PI 3-kinase p110 complex. Expressed in heart, dendritic cells, macrophages and neutrophils, p87PIKAP drives PI 3-kinase p110 activation, interacts with PDE3B and is thought to cause the two proteins to form a scaffolding interaction. The gene encoding p87PIKAP maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1, 200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Specifications
PIK3R6 | |
Polyclonal | |
Unconjugated | |
PIK3R6 | |
BB220380; C17orf38; HsT41028; p84; p84 PI3K adapter protein; p84 PIKAP; p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein; p87 phosphoinositide 3-kinase gamma adapter; p87 PI3K adapter protein; p87(PIKAP); p87PIKAP; Phosphoinositide 3-kinase gamma adapter protein of 87 kDa; Phosphoinositide 3-kinase regulatory subunit 6; phosphoinositide-3-kinase regulatory subunit 6; phosphoinositide-3-kinase, regulatory subunit 6; PI3Kgamma adapter protein of 87 kDa; PIK3R6; RGD1560850 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146850 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5UE93 | |
PIK3R6 | |
Recombinant protein corresponding to Human PIK3R6. Recombinant protein control fragment (Product #RP-93428). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction