Learn More
Pikachurin Mouse anti-Human, Clone: 1C6D12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Pikachurin |
|---|---|
| Clone | 1C6D12 |
| Concentration | 2 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6665
![]()
|
Proteintech Group Inc
662321IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This monoclonal antibody detects two bands of 111 kDa and 55-60 kDa. The lower-molecular-weight band has been reported and likely represents a fragment resulting from cleavage of Pikachurin.
Involved in both the retinal photoreceptor ribbon synapse formation and physiological functions of visual perception. Necessary for proper bipolar dendritic tip apposition to the photoreceptor ribbon synapse. Promotes matrix assembly and cell adhesiveness.Specifications
| Pikachurin | |
| 2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Agrin like protein, AGRINL, AGRNL, EGFLAM, Pikachurin | |
| EGFLAM | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1C6D12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q63HQ2 | |
| 133584 | |
| Pikachurin Fusion Protein Ag6362 | |
| Primary | |
| -20°C | |
| EGFLAM |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.