Learn More
Pikachurin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | Pikachurin |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9557
![]()
|
Proteintech Group Inc
145781AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody detects a 60- kDa band, which has been reported and likely represents a fragment resulting from cleavage of Pikachurin.
Involved in both the retinal photoreceptor ribbon synapse formation and physiological functions of visual perception. Necessary for proper bipolar dendritic tip apposition to the photoreceptor ribbon synapse. Promotes matrix assembly and cell adhesiveness.Specifications
| Pikachurin | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| B4F785, Q4VBE4, Q63HQ2 | |
| 133584, 268780, 365691 | |
| Pikachurin Fusion Protein Ag5986 | |
| Primary | |
| -20°C | |
| EGFLAM |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Agrin like protein, AGRINL, AGRNL, EGFLAM, Pikachurin | |
| Egflam | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.