Learn More
PIKFYVE Monoclonal Antibody (OTI1D2)

Description
PIKFYVE Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PIKFYVE |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PIKFYVE |
| Gene Accession No. | Q9Y2I7 |
| Gene Alias | 1-phosphatidylinositol 3-phosphate 5-kinase; 1-phosphatidylinositol-3-phosphate 5-kinase; 5230400C17Rik; CFD; epididymis luminal protein 37; FAB1; FYVE finger-containing phosphoinositide kinase; HEL37; Kiaa0981; p235; Phosphatidylinositol 3-phosphate 5-kinase; Phosphatidylinositol 3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate/phosphatidylinositol 5-kinase, type III; phosphatidylinositol-4-phosphate 5-kinase, type III; phosphoinositide kinase, FYVE finger containing; phosphoinositide kinase, FYVE-type zinc finger containing; PIKfyve; Pip5k; PIP5K3; Pipk5k3; PipkIII; PIPkin-III; PtdIns(4)P-5-kinase; type III PIP kinase; ZFYVE29; zinc finger, FYVE domain containing 29 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.